5-(Hexamethyleneimin-1-yl)-2-methylsulfonylaniline

Supplier: Matrix Scientific

024491-500MG
101818-188EA 267.69 USD
101818-188
5-(Hexamethyleneimin-1-yl)-2-methylsulfonylaniline
5-(Hexamethyleneimin-1-yl)-2-methylsulfonylaniline

Hazard: Irritant

Formula: C₁₃H₂₀N₂O₂S
MDL Number: MFCD09038303
CAS Number: 942474-92-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR