Ethyl-6-chloroimidazo[1,2-b]pyridazine-2-carboxylate

Supplier: Matrix Scientific

Synonyms: Ethyl 6-chloroimidazo[1,2-b]pyridazine-2-carboxylate

024481-500MG
101818-168EA 56.92 USD
101818-168
Ethyl-6-chloroimidazo[1,2-b]pyridazine-2-carboxylate
Ethyl 6-chloroimidazo[1,2-b]pyridazine-2-carboxylate

Hazard: Irritant

Formula: C₉H₈ClN₃O₂
MW: 225.6317 g/mol
MDL Number: MFCD03086230
CAS Number: 64067-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR