3-Nitro-4-(pyrrolidin-1-yl)benzoic acid

Supplier: Matrix Scientific

024475-1G
101818-160EA 218.46 USD
101818-160
3-Nitro-4-(pyrrolidin-1-yl)benzoic acid
3-Nitro-4-(pyrrolidin-1-yl)benzoic acid

Hazard: Irritant

Formula: C₁₁H₁₂N₂O₄
MDL Number: MFCD02112810
CAS Number: 40832-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR