Ethyl-1-[2-(methylsulfonyl)-4-nitrophenyl]piperidine-4-carboxalate

Supplier: Matrix Scientific

024469-100MG
101818-148EA 218.46 USD
101818-148
Ethyl-1-[2-(methylsulfonyl)-4-nitrophenyl]piperidine-4-carboxalate
Ethyl-1-[2-(methylsulfonyl)-4-nitrophenyl]piperidine-4-carboxalate

Hazard: Irritant

Formula: C₁₅H₂₀N₂O₆S
MDL Number: MFCD08704871
CAS Number: 951625-05-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR