2-Benzenesulfonyl-4-methoxybenzaldehyde

Supplier: Matrix Scientific

024467-500MG
101818-144EA 306.15 USD
101818-144
2-Benzenesulfonyl-4-methoxybenzaldehyde
2-Benzenesulfonyl-4-methoxybenzaldehyde

Hazard: Irritant

Formula: C₁₄H₁₂O₄S
MDL Number: MFCD08692479
CAS Number: 942474-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR