1-[4-Fluoro-2-(methylsulfonyl)phenyl]hydrazine

Supplier: Matrix Scientific

024466-250MG
101818-142EA 175.38 USD
101818-142
1-[4-Fluoro-2-(methylsulfonyl)phenyl]hydrazine
1-[4-Fluoro-2-(methylsulfonyl)phenyl]hydrazine

Hazard: Irritant

Formula: C₇H₉FN₂O₂S
MW: 204.22 g/mol
MDL Number: MFCD08692478
CAS Number: 914637-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR