1-[4-Fluoro-2-(methylsulfonyl)phenyl]homopiperazine hydrochloride

Supplier: Matrix Scientific

024461-100MG
101818-132EA 267.69 USD
101818-132
1-[4-Fluoro-2-(methylsulfonyl)phenyl]homopiperazine hydrochloride
1-[4-Fluoro-2-(methylsulfonyl)phenyl]homopiperazine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₈ClFN₂O₂S
MDL Number: MFCD08692474
CAS Number: 1185015-65-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR