1-[4-Fluoro-2-(methylsulfonyl)phenyl]piperidine-3-carboxylic acid

Supplier: Matrix Scientific

024456-100MG
101818-122EA 267.69 USD
101818-122
1-[4-Fluoro-2-(methylsulfonyl)phenyl]piperidine-3-carboxylic acid
1-[4-Fluoro-2-(methylsulfonyl)phenyl]piperidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₆FNO₄S
MDL Number: MFCD08692470
CAS Number: 914637-71-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR