1-(2-Nitrophenyl)piperidine-4-carboxylic acid

Supplier: Matrix Scientific

024455-500MG
101818-120EA 241.54 USD
101818-120
1-(2-Nitrophenyl)piperidine-4-carboxylic acid
1-(2-Nitrophenyl)piperidine-4-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₄N₂O₄
MDL Number: MFCD06362460
CAS Number: 438192-02-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR