1-[4-Fluoro-2-(methylsulfonyl)phenyl]piperidine-4-carboxamide

Supplier: Matrix Scientific

024454-250MG
101818-118EA 472.31 USD
101818-118
1-[4-Fluoro-2-(methylsulfonyl)phenyl]piperidine-4-carboxamide
1-[4-Fluoro-2-(methylsulfonyl)phenyl]piperidine-4-carboxamide

Hazard: Irritant

Formula: C₁₃H₁₇FN₂O₃S
MDL Number: MFCD08692469
CAS Number: 914637-65-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR