2-[4-[5-(Trifluoromethyl)pyridin-2-yl]piperazin-1-yl]ethanol

Supplier: Matrix Scientific

Synonyms: 1-(2-Hydroxyethyl)-4-[5-(trifluoromethyl)pyridin-2-yl]piperazine

024440-250MG
101818-096EA 153.85 USD
101818-096
2-[4-[5-(Trifluoromethyl)pyridin-2-yl]piperazin-1-yl]ethanol
2-[4-[5-(Trifluoromethyl)pyridin-2-yl]piperazin-1-yl]ethanol

Hazard: Irritant

Formula: C₁₂H₁₆F₃N₃O
MW: 275.28 g/mol
MDL Number: MFCD00203918
CAS Number: 215434-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR