1-(3-Nitropyridin-2-yl)piperidine-3-carboxylic acid

Supplier: Matrix Scientific

024432-500MG
101818-082EA 447.69 USD
101818-082
1-(3-Nitropyridin-2-yl)piperidine-3-carboxylic acid
1-(3-Nitropyridin-2-yl)piperidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₃N₃O₄
MDL Number: MFCD08692459
CAS Number: 937606-75-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR