1-(4-Fluoro-2-nitrophenyl)homopiperazine hydrochloride

Supplier: Matrix Scientific

024428-250MG
101818-076EA 447.69 USD
101818-076
1-(4-Fluoro-2-nitrophenyl)homopiperazine hydrochloride
1-(4-Fluoro-2-nitrophenyl)homopiperazine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₅ClFN₃O₂
MDL Number: MFCD08692456
CAS Number: 1185298-75-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR