1-(5-Nitropyridin-2-yl)-3-piperidinecarboxylic acid

Supplier: Matrix Scientific

024426-250MG
101818-072EA 253.85 USD
101818-072
1-(5-Nitropyridin-2-yl)-3-piperidinecarboxylic acid
1-(5-Nitropyridin-2-yl)-3-piperidinecarboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₃N₃O₄
MDL Number: MFCD08692455
CAS Number: 937606-76-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR