1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-4-piperidinecarboxylic acid

Supplier: Matrix Scientific

024415-250MG
101818-050EA 153.85 USD
101818-050
1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-4-piperidinecarboxylic acid
1-[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]-4-piperidinecarboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₂ClF₃N₂O₂
MW: 308.69 g/mol
MDL Number: MFCD00172245
CAS Number: 337919-65-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR