1-(4-Fluoro-2-nitrophenyl)-3-methylpiperidine

Supplier: Matrix Scientific

024408-500MG
101818-036EA 192.31 USD
101818-036
1-(4-Fluoro-2-nitrophenyl)-3-methylpiperidine
1-(4-Fluoro-2-nitrophenyl)-3-methylpiperidine

Hazard: Irritant

Formula: C₁₂H₁₅FN₂O₂
MDL Number: MFCD08692445
CAS Number: 942474-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR