5-Fluoro-2-piperidin-1-ylaniline hydrochloride

024406-250MG
101818-032EA 445.83 USD
101818-032
5-Fluoro-2-piperidin-1-ylaniline hydrochloride
5-Fluoro-2-piperidin-1-ylaniline hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₆ClFN₂
MDL Number: MFCD08692443
CAS Number: 1185302-55-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR