4-(4-Fluoro-2-nitrophenyl)morpholine

Supplier: Matrix Scientific

024403-1G
101818-026EA 315.38 USD
101818-026
4-(4-Fluoro-2-nitrophenyl)morpholine
4-(4-Fluoro-2-nitrophenyl)morpholine

Hazard: Irritant

Formula: C₁₀H₁₁FN₂O₃
MDL Number: MFCD00798369
CAS Number: 238418-75-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR