1-(4-Fluoro-2-nitrophenyl)piperidine-4-carboxylic acid
Supplier: Matrix Scientific
024399-250MG
101818-018EA
160
USD
101818-018
1-(4-Fluoro-2-nitrophenyl)piperidine-4-carboxylic acid
1-(4-Fluoro-2-nitrophenyl)piperidine-4-carboxylic acid
Hazard: Irritant
Formula:
C₁₂H₁₃FN₂O₄ |
MDL Number:
MFCD05664786 CAS Number: 648917-79-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...