1-(2-Nitrophenyl)piperidine-3-carboxylic acid

Supplier: Matrix Scientific

024397-500MG
101818-014EA 375.38 USD
101818-014
1-(2-Nitrophenyl)piperidine-3-carboxylic acid
1-(2-Nitrophenyl)piperidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₄N₂O₄
MDL Number: MFCD08442646
CAS Number: 926224-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR