2-[4-[2-(Methylsulfonyl)phenyl]piperazin-1-yl]-ethanol

Supplier: Matrix Scientific

024393-250MG
101818-006EA 235.38 USD
101818-006
2-[4-[2-(Methylsulfonyl)phenyl]piperazin-1-yl]-ethanol
2-[4-[2-(Methylsulfonyl)phenyl]piperazin-1-yl]-ethanol

Hazard: Irritant

Formula: C₁₃H₂₀N₂O₃S
MDL Number: MFCD08692437
CAS Number: 942474-21-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR