3-Nitro-4-piperidin-1-ylbenzoic acid

Supplier: Matrix Scientific

024382-500MG
101817-986EA 203.08 USD
101817-986
3-Nitro-4-piperidin-1-ylbenzoic acid
3-Nitro-4-piperidin-1-ylbenzoic acid

Hazard: Irritant

Formula: C₁₂H₁₄N₂O₄
MDL Number: MFCD00157001
CAS Number: 26586-26-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR