1-(3-Methyl-4-nitrophenyl)piperidine-2-carboxylic acid

Supplier: Matrix Scientific

024381-250MG
101817-984EA 283.08 USD
101817-984
1-(3-Methyl-4-nitrophenyl)piperidine-2-carboxylic acid
1-(3-Methyl-4-nitrophenyl)piperidine-2-carboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₆N₂O₄
MDL Number: MFCD08692432
CAS Number: 1189547-44-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR