1-[2-Fluoro-4-(methylsulfonyl)phenyl]homopiperazine hydrochloride

Supplier: Matrix Scientific

024376-100MG
101817-976EA 267.69 USD
101817-976
1-[2-Fluoro-4-(methylsulfonyl)phenyl]homopiperazine hydrochloride
1-[2-Fluoro-4-(methylsulfonyl)phenyl]homopiperazine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₈ClFN₂O₂S
MDL Number: MFCD08692428
CAS Number: 1185298-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR