1-[2-Fluoro-4-(methylsulfonyl)phenyl]piperidine-4-carboxamide

Supplier: Matrix Scientific

024375-250MG
101817-974EA 320 USD
101817-974
1-[2-Fluoro-4-(methylsulfonyl)phenyl]piperidine-4-carboxamide
1-[2-Fluoro-4-(methylsulfonyl)phenyl]piperidine-4-carboxamide

Hazard: Irritant

Formula: C₁₃H₁₇FN₂O₃S
MDL Number: MFCD08692427
CAS Number: 914637-63-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR