1-[4-(Methylsulfonyl)phenyl]piperidine-4-carboxamide

Supplier: Matrix Scientific

024367-250MG
101817-958EA 235.38 USD
101817-958
1-[4-(Methylsulfonyl)phenyl]piperidine-4-carboxamide
1-[4-(Methylsulfonyl)phenyl]piperidine-4-carboxamide

Hazard: Irritant

Formula: C₁₃H₁₈N₂O₃S
MDL Number: MFCD08692420
CAS Number: 942474-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR