1-[4-(Methylsulfonyl)phenyl]piperidine-4-carboxylic acid

Supplier: Matrix Scientific

024366-250MG
101817-956EA 283.08 USD
101817-956
1-[4-(Methylsulfonyl)phenyl]piperidine-4-carboxylic acid
1-[4-(Methylsulfonyl)phenyl]piperidine-4-carboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₇NO₄S
MDL Number: MFCD08692419
CAS Number: 951624-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR