1-(3-Methyl-4-nitrophenyl)piperidine-3-carboxylic acid

Supplier: Matrix Scientific

024362-250MG
101817-948EA 253.85 USD
101817-948
1-(3-Methyl-4-nitrophenyl)piperidine-3-carboxylic acid
1-(3-Methyl-4-nitrophenyl)piperidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₆N₂O₄
MDL Number: MFCD08692415
CAS Number: 942474-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR