1-(4-Chloro-2-nitrophenyl)piperidine-4-carboxylic acid

Supplier: Matrix Scientific

024360-250MG
101817-944EA 235.38 USD
101817-944
1-(4-Chloro-2-nitrophenyl)piperidine-4-carboxylic acid
1-(4-Chloro-2-nitrophenyl)piperidine-4-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₃ClN₂O₄
MDL Number: MFCD05664784
CAS Number: 874800-64-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR