1-[4-Nitro-3-(trifluoromethyl)phenyl]homopiperazine hydrochloride

Supplier: Matrix Scientific

024357-100MG
101817-938EA 267.69 USD
101817-938
1-[4-Nitro-3-(trifluoromethyl)phenyl]homopiperazine hydrochloride
1-[4-Nitro-3-(trifluoromethyl)phenyl]homopiperazine hydrochloride

Hazard: Irritant

Formula: C₁₂H₁₅ClF₃N₃O₂
MDL Number: MFCD09025926
CAS Number: 1185298-60-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR