3-Methyl-1-(4-nitrophenyl)piperazine hydrochloride

Supplier: Matrix Scientific

024355-500MG
101817-934EA 278.46 USD
101817-934
3-Methyl-1-(4-nitrophenyl)piperazine hydrochloride
3-Methyl-1-(4-nitrophenyl)piperazine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₆ClN₃O₂
MDL Number: MFCD00545967
CAS Number: 1158439-97-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR