1-(4-Nitrophenyl)-3-piperidinecarboxylic acid

Supplier: Matrix Scientific

024353-250MG
101817-930EA 330.77 USD
101817-930
1-(4-Nitrophenyl)-3-piperidinecarboxylic acid
1-(4-Nitrophenyl)-3-piperidinecarboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₄N₂O₄
CAS Number: 926224-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR