1-(3-Methyl-4-nitrophenyl)piperazine

Supplier: Matrix Scientific

024352-500MG
101817-928EA 241.54 USD
101817-928
1-(3-Methyl-4-nitrophenyl)piperazine
1-(3-Methyl-4-nitrophenyl)piperazine

Hazard: Irritant

Formula: C₁₁H₁₅N₃O₂
MDL Number: MFCD08692413
CAS Number: 192441-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR