1-(2-Fluoro-4-nitrophenyl)piperidine-2-carboxylic acid

Supplier: Matrix Scientific

024351-100MG
101817-926EA 200.1 USD
101817-926
1-(2-Fluoro-4-nitrophenyl)piperidine-2-carboxylic acid
1-(2-Fluoro-4-nitrophenyl)piperidine-2-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₃FN₂O₄
CAS Number: 1214189-74-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR