1-(4-Chloro-2-nitrophenyl)-3-methylpiperazine hydrochloride

Supplier: Matrix Scientific

024348-500MG
101817-922EA 278.46 USD
101817-922
1-(4-Chloro-2-nitrophenyl)-3-methylpiperazine hydrochloride
1-(4-Chloro-2-nitrophenyl)-3-methylpiperazine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₅Cl₂N₃O₂
MDL Number: MFCD00545970
CAS Number: 331657-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR