1-(2-Chloro-4-nitrophenyl)-3-methylpiperazine hydrochloride

Supplier: Matrix Scientific

024347-500MG
101817-920EA 278.46 USD
101817-920
1-(2-Chloro-4-nitrophenyl)-3-methylpiperazine hydrochloride
1-(2-Chloro-4-nitrophenyl)-3-methylpiperazine hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₅Cl₂N₃O₂
MDL Number: MFCD00545969
CAS Number: 392710-17-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR