1-(2-Fluoro-4-nitrophenyl)-3-methylpiperazine

Supplier: Matrix Scientific

024345-250MG
101817-916EA 224.62 USD
101817-916
1-(2-Fluoro-4-nitrophenyl)-3-methylpiperazine
1-(2-Fluoro-4-nitrophenyl)-3-methylpiperazine

Hazard: Irritant

Formula: C₁₁H₁₄FN₃O₂
MDL Number: MFCD08692408
CAS Number: 837421-28-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR