1-(2-Fluoro-4-nitrophenyl)piperidine-4-carboxylic acid

Supplier: Matrix Scientific

024344-250MG
101817-914EA 283.08 USD
101817-914
1-(2-Fluoro-4-nitrophenyl)piperidine-4-carboxylic acid
1-(2-Fluoro-4-nitrophenyl)piperidine-4-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₃FN₂O₄
MW: 268.24 g/mol
MDL Number: MFCD08692407
CAS Number: 915187-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR