1-[4-(Ethylsulfonyl)-2-nitrophenyl]-4-methylpiperidine

Supplier: Matrix Scientific

024342-250MG
101817-910EA 224.62 USD
101817-910
1-[4-(Ethylsulfonyl)-2-nitrophenyl]-4-methylpiperidine
1-[4-(Ethylsulfonyl)-2-nitrophenyl]-4-methylpiperidine

Hazard: Irritant

Formula: C₁₄H₂₀N₂O₄S
MDL Number: MFCD08692406
CAS Number: 942474-76-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR