1-[4-(Ethylsulfonyl)-2-nitrophenyl]hexamethyleneimine

Supplier: Matrix Scientific

024339-250MG
101817-904EA 224.62 USD
101817-904
1-[4-(Ethylsulfonyl)-2-nitrophenyl]hexamethyleneimine
1-[4-(Ethylsulfonyl)-2-nitrophenyl]hexamethyleneimine

Hazard: Irritant

Formula: C₁₄H₂₀N₂O₄S
MDL Number: MFCD08704868
CAS Number: 942474-69-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR