1-[4-(Ethylsulfonyl)-2-nitrophenyl]piperidine

Supplier: Matrix Scientific

024337-250MG
101817-900EA 224.62 USD
101817-900
1-[4-(Ethylsulfonyl)-2-nitrophenyl]piperidine
1-[4-(Ethylsulfonyl)-2-nitrophenyl]piperidine

Hazard: Irritant

Formula: C₁₃H₁₈N₂O₄S
MDL Number: MFCD08692403
CAS Number: 942474-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR