1-[4-(Ethylsulfonyl)-2-nitrophenyl]pyrrolidine-2-carboxylic acid

Supplier: Matrix Scientific

024336-250MG
101817-898EA 320 USD
101817-898
1-[4-(Ethylsulfonyl)-2-nitrophenyl]pyrrolidine-2-carboxylic acid
1-[4-(Ethylsulfonyl)-2-nitrophenyl]pyrrolidine-2-carboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₆N₂O₆S
MDL Number: MFCD08692402
CAS Number: 1452575-76-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR