Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Bromo-4-(ethylsulphonyl)-2-nitrobenzene

Supplier: Matrix Scientific


024334-500MG
101817-894EA 340 USD InStock
101817-894
1-Bromo-4-(ethylsulphonyl)-2-nitrobenzene
1-Bromo-4-(ethylsulphonyl)-2-nitrobenzene
CAS: 942474-83-1
MDL Number: MFCD08692401
Molecular Formula: C8H8BrNO4S
Molecular Weight: 294,13
Hazard: Irritant


Formula: C₈H₈BrNO₄S
CAS Number: 942474-83-1
EINECS: 000-000-0
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty