8-[4-(Ethylsulfonyl)-2-nitrophenyl]-1,4-dioxa-8-azaspiro[4.5]decane

Supplier: Matrix Scientific

024331-250MG
101817-888EA 273.85 USD
101817-888
8-[4-(Ethylsulfonyl)-2-nitrophenyl]-1,4-dioxa-8-azaspiro[4.5]decane
8-[4-(Ethylsulfonyl)-2-nitrophenyl]-1,4-dioxa-8-azaspiro[4.5]decane

Hazard: Irritant

Formula: C₁₅H₂₀N₂O₆S
MDL Number: MFCD08692399
CAS Number: 942474-61-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR