4-(Ethylsulfonyl)-2-nitroanisole

Supplier: Matrix Scientific

024327-250MG
101817-880EA 206.15 USD
101817-880
4-(Ethylsulfonyl)-2-nitroanisole
4-(Ethylsulphonyl)-2-nitroanisole

Hazard: Irritant

Formula: C₉H₁₁NO₅S
MDL Number: MFCD08692396
CAS Number: 51572-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR