1-Bromo-4-(ethylsulfonyl)benzene

Supplier: Matrix Scientific

024313-500MG
101817-854EA 118.46 USD
101817-854
1-Bromo-4-(ethylsulfonyl)benzene
1-Bromo-4-(ethylsulfonyl)benzene

Hazard: Irritant

Formula: C₈H₉BrO₂S
MDL Number: MFCD08692383
CAS Number: 26732-20-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR