1-(2-Fluoro-4-nitrophenyl)piperidine-4-carboxamide

Supplier: Matrix Scientific

024310-250MG
101817-848EA 224.62 USD
101817-848
1-(2-Fluoro-4-nitrophenyl)piperidine-4-carboxamide
1-(2-Fluoro-4-nitrophenyl)piperidine-4-carboxamide

Hazard: Irritant

Formula: C₁₂H₁₄FN₃O₃
MDL Number: MFCD08692380
CAS Number: 912894-29-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR