1-(2-Fluoro-4-nitrophenyl)homopiperazine

Supplier: Matrix Scientific

024309-500MG
101817-846EA 118.46 USD
101817-846
1-(2-Fluoro-4-nitrophenyl)homopiperazine
1-(2-Fluoro-4-nitrophenyl)homopiperazine

Hazard: Irritant

Formula: C₁₁H₁₄FN₃O₂
MDL Number: MFCD08692379
CAS Number: 392660-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR