4-Amino-N-butylbenzamide

Supplier: Matrix Scientific

024297-500MG
101817-814EA 133.85 USD
101817-814
4-Amino-N-butylbenzamide
4-Amino-N-butylbenzamide

Hazard: Irritant

Formula: C₁₁H₁₆N₂O
MDL Number: MFCD01940422
CAS Number: 51207-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR