3-Amino-N,N-diethylbenzamide

Supplier: Matrix Scientific

024296-500MG
101817-812EA 78.46 USD
101817-812
3-Amino-N,N-diethylbenzamide
3-Amino-N,N-diethylbenzamide

Hazard: Irritant

Formula: C₁₁H₁₆N₂O
MDL Number: MFCD00025279
CAS Number: 68269-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR